Skip to Content
Merck
All Photos(2)

Documents

332550

Sigma-Aldrich

N,N,N′-Trimethyl-1,3-propanediamine

96%

Synonym(s):

(3-Dimethylaminopropyl)methylamine, 3-(Dimethylamino)-N-methylpropylamine, Methyl[3-(dimethylamino)propyl]amine, N,N,N′-Trimethyl-1,3-diaminopropane, N,N,N′-Trimethylpropan-1,3-diamine, N,N,N′-Trimethyltrimethylenediamine, N-(3-Dimethylaminopropyl)methylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2N(CH2)3NHCH3
CAS Number:
Molecular Weight:
116.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.4283 (lit.)

bp

140-142 °C (lit.)

density

0.793 g/mL at 25 °C (lit.)

SMILES string

CNCCCN(C)C

InChI

1S/C6H16N2/c1-7-5-4-6-8(2)3/h7H,4-6H2,1-3H3

InChI key

SORARJZLMNRBAQ-UHFFFAOYSA-N

Application

N,N,N′-Trimethyl-1,3-propanediamine was used in preparation of unlabelled N,N,N′-trimethyl-[2-hydroxy-3-methyl-5-iodobenzyl]-1,3-propanediamine (HIDPM).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B Lui et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 28(3), 360-365 (1987-03-01)
In conjunction with single photon emission computed tomography (SPECT), iodine-123 (123I)-labeled N,N,N'-trimethyl-[2-hydroxy-3-methyl-5-iodobenzyl]-1,3-propanediamine (HIPDM) has been used clinically as a regional cerebral perfusion imaging agent. The [123I]HIPDM can be prepared by a simple aqueous exchange reaction in a kit form. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service