All Photos(1)
About This Item
Linear Formula:
ClC6H3(OH)CHO
CAS Number:
Molecular Weight:
156.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
145-147 °C (lit.)
functional group
aldehyde
chloro
SMILES string
Oc1ccc(C=O)c(Cl)c1
InChI
1S/C7H5ClO2/c8-7-3-6(10)2-1-5(7)4-9/h1-4,10H
InChI key
ZMOMCILMBYEGLD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
N-Thioamide thiosemicarbazone derivatives of 2-chloro-4-hydroxy-benzaldehyde have been prepared. pK values for acid dissociation of 2-chloro-4-hydroxybenzaldehyde has been evaluated.
Application
2-Chloro-4-hydroxybenzaldehyde may be used in chemical synthesis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Coordination in Solutions. II. Acid Dissociation Constants in Water and Structural Reassignments of the Isomeric Chlorosalicylaldehydes1, 2.
Postmus Jr C, et al.
The Journal of Organic Chemistry, 29(9), 2693-2698 (1964)
Synthesis and characterization of thiosemicarbazone derivatives of 2-chloro-4-hydroxy-benzaldehyde and their rhenium (I) complexes.
Nu?ez-Montenegro A, et al.
Polyhedron, 28(18), 3915-3922 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service