Skip to Content
Merck
All Photos(1)

Key Documents

340456

Sigma-Aldrich

Pyrazine-d4

98 atom % D

Synonym(s):

Deuterated pyrazine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€333.00
5 G
€1,220.00

€333.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€333.00
5 G
€1,220.00

About This Item

Empirical Formula (Hill Notation):
C4D4N2
CAS Number:
Molecular Weight:
84.11
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€333.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

bp

116 °C (lit.)

mp

55-57 °C (lit.)

mass shift

M+4

SMILES string

[2H]c1nc([2H])c([2H])nc1[2H]

InChI

1S/C4H4N2/c1-2-6-4-3-5-1/h1-4H/i1D,2D,3D,4D

InChI key

KYQCOXFCLRTKLS-RHQRLBAQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rohit Mahar et al.
Scientific reports, 10(1), 8885-8885 (2020-06-03)
Increased glucose uptake and aerobic glycolysis are striking features of many cancers. These features have led to many techniques for screening and diagnosis, but many are expensive, less feasible or have harmful side-effects. Here, we report a sensitive 1H/2H NMR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service