Skip to Content
Merck
All Photos(4)

Documents

339873

Sigma-Aldrich

Diiodosilane

greener alternative

contains copper as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiH2I2
CAS Number:
Molecular Weight:
283.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

contains

copper as stabilizer

greener alternative product score

old score: 3
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

56-60 °C/25 mmHg (lit.)

density

2.834 g/mL at 25 °C (lit.)

greener alternative category

storage temp.

2-8°C

SMILES string

I[Si]([H])([H])I

InChI

1S/H2I2Si/c1-3-2/h3H2

InChI key

AIHCVGFMFDEUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Safer Solvents and Auxiliaries” and “Atom Economy”. Click here to view its DOZN scorecard.

Application

Reagent involved in reductive iodination for synthesis of [18F]FDOPA

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service