Skip to Content
Merck
All Photos(1)

Key Documents

339709

Sigma-Aldrich

Nickel(II) hexafluoroacetylacetonate hydrate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€90.30

€90.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€90.30

About This Item

Linear Formula:
Ni(C5HF6O2)2 · xH2O
CAS Number:
Molecular Weight:
472.79 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€90.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

core: nickel
reagent type: catalyst

mp

213 °C (dec.) (lit.)

SMILES string

O.FC(F)(F)C(=O)\C=C(/O[Ni]O\C(=C/C(=O)C(F)(F)F)C(F)(F)F)C(F)(F)F

InChI

1S/2C5H2F6O2.Ni.H2O/c2*6-4(7,8)2(12)1-3(13)5(9,10)11;;/h2*1,12H;;1H2/q;;+2;/p-2/b2*2-1-;;

InChI key

OMCOVIODBHSWJG-SUXDNRKISA-L

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service