Skip to Content
Merck
All Photos(2)

Key Documents

339695

Sigma-Aldrich

Cobalt(II) hexafluoroacetylacetonate hydrate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€118.00

€118.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€118.00

About This Item

Linear Formula:
Co(C5HF6O2)2 · xH2O
CAS Number:
Molecular Weight:
473.03 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€118.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder or crystals
solid

composition

Degree of hydration, 2-3

reaction suitability

core: cobalt

mp

197 °C (dec.) (lit.)

SMILES string

O.FC(F)(F)C(=O)\C=C(/O[Co]O\C(=C/C(=O)C(F)(F)F)C(F)(F)F)C(F)(F)F

InChI

1S/2C5H2F6O2.Co.H2O/c2*6-4(7,8)2(12)1-3(13)5(9,10)11;;/h2*1,12H;;1H2/q;;+2;/p-2/b2*2-1-;;

InChI key

XPVNKVXSBTZRRC-SUXDNRKISA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service