Skip to Content
Merck
All Photos(1)

Key Documents

228575

Sigma-Aldrich

4-Hydroxy-3-nitrobenzoic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€70.30

€70.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€70.30

About This Item

Linear Formula:
HOC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
183.12
Beilstein:
2213134
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€70.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

183-186 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1ccc(O)c(c1)[N+]([O-])=O

InChI

1S/C7H5NO5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H,(H,10,11)

InChI key

QRYSWXFQLFLJTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxy-3-nitrobenzoic acid was coupled to ionic liquid-immobilized o-phenylenediamine to form benzimidazole derivatives[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaushik Chanda et al.
ACS combinatorial science, 14(2), 115-123 (2012-01-24)
A novel and efficient diversity-oriented synthetic approach was employed to access the benzo[d]oxazol-5-yl-1H-benzo[d]imidazole on ionic liquid support, which helps to absorb microwave irradiation. In this paper, we successfully coupled 4-hydroxy-3-nitrobenzoic acid onto ionic liquid-immobilized o-phenylenediamine, which subsequently underwent an acid
Yuefei Ji et al.
Water research, 123, 249-257 (2017-07-04)
As promising in-situ chemical oxidation (ISCO) technologies, sulfate radical-based advanced oxidation processes (SR-AOPs) are applied in wastewater treatment and groundwater remediation in recent years. In this contribution, we report for the first time that, thermally activated persulfate oxidation of phenol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service