Skip to Content
Merck
All Photos(1)

Key Documents

338761

Sigma-Aldrich

Triphenylphosphine-d15

≥98 atom % D, 99% (CP)

Synonym(s):

Tris(pentadeuterophenyl)phosphine, Tri(phenyl-d5)phosphine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€282.00

€282.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€282.00

About This Item

Linear Formula:
(C6D5)3P
CAS Number:
Molecular Weight:
277.38
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.12

€282.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Assay

99% (CP)

form

powder

bp

377 °C

mp

79-81 °C (lit.)

mass shift

M+15

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])P(c2c([2H])c([2H])c([2H])c([2H])c2[2H])c3c([2H])c([2H])c([2H])c([2H])c3[2H]

InChI

1S/C18H15P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D,15D

InChI key

RIOQSEWOXXDEQQ-KLHTYYPYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in a mechanistic study of cis-bis(silylmethyl)platinum(II) complexes.[1]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1B - STOT RE 1 Inhalation

Target Organs

Central nervous system,Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 116, 9963-9963 (1994)
Elliot L Bennett et al.
Inorganic chemistry, 53(6), 2963-2967 (2014-02-28)
A structural, spectroscopic, and computational investigation of Stryker's reagent, [HCu{P(C6H5)3}]6, and its isotopomers has provided new insights into the complex. Neutron diffraction shows that the hydrides are best described as edge bridging rather than face bridging. The combination of infrared
Elliot Bennett et al.
Inorganic chemistry, 54(5), 2213-2220 (2015-02-12)
CuH is a material that appears in a wide diversity of circumstances ranging from catalysis to electrochemistry to organic synthesis. There are both aqueous and nonaqueous synthetic routes to CuH, each of which apparently leads to a different product. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service