Skip to Content
Merck
All Photos(1)

Key Documents

338680

Sigma-Aldrich

4′-Octyl-4-biphenylcarbonitrile

liquid crystal (nematic), 98%

Synonym(s):

4′-n -Octyl-4-cyanobiphenyl, 4′-Octyl-4-biphenylcarbonitrile, 4′-Octyl-4-cyanobiphenyl, 4′-Octyl[1,1′-biphenyl]-4-carbonitrile, 4-n -Octyl-4′-cyanobiphenyl, 4-Cyano-4′-n -octylbiphenyl, 4-Cyano-4′-octyl-1,1′-biphenyl, 4-Cyano-4′-octylbiphenyl, 4-Octyl-4′-cyanobiphenyl, p -n -Octyl-p ′-cyanobiphenyl, p -Cyano-p ′-octylbiphenyl, p -Octyl-p ′-cyanobiphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€136.00

€136.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€136.00

About This Item

Linear Formula:
CH3(CH2)7C6H4C6H4CN
CAS Number:
Molecular Weight:
291.43
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€136.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid crystal (nematic)

SMILES string

CCCCCCCCc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C21H25N/c1-2-3-4-5-6-7-8-18-9-13-20(14-10-18)21-15-11-19(17-22)12-16-21/h9-16H,2-8H2,1H3

InChI key

CSQPODPWWMOTIY-UHFFFAOYSA-N

General description

4′-octyl-4-biphenylcarbonitrile (8CB) is a low molar mass liquid crystal (LC), which shows different liquid crystalline phases depending on temperature conditions. Under normal temperature conditions 8CB exists only as a bulk smectic-A phase.[1] LCs are responsive to subtle changes in temperature, shear, electric (or magnetic) fields, and the structure of solid surfaces with which they are in contact.[2]

Application

Monolayer films of the LCs deposited onto highly ordered pyrolytic graphite, these self-assembled films display a wide variety of structural forms.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monirosadat Sadati et al.
Journal of the American Chemical Society, 139(10), 3841-3850 (2017-02-09)
Numerous applications of liquid crystals rely on control of molecular orientation at an interface. However, little is known about the precise molecular structure of such interfaces. In this work, synchrotron X-ray reflectivity measurements, accompanied by large-scale atomistic molecular dynamics simulations
Substrate defects and variations in interfacial ordering of monolayer molecular films on graphite.
Patrick DL, et al.
Langmuir, 10(1), 298-302 (1994)
Mechanism of molecular ordering in monolayer liquid crystal films.
Patrick DL, et al.
The Journal of Physical Chemistry, 100(20), 8478-8481 (1996)
Separation of nematic and smectic-A potential effects on solute ordering in a series of binary 6OCB-8OCB mixtures
Burnell EE, et al.
Canadian Journal of Chemistry, 89(7), 900-908 (2011)

Questions

1–3 of 3 Questions  
  1. Give the phase transition temperatures and name the different phases of this liquid crystalline compound 338680 4'-octyl-4-biphenyl carbonitrile

    1 answer
    1. The phase transition temperatures for this compound have not been determined. See the link below to the PubChem file for this material. There are a number of references and physical properties listed that may be of interest.
      https://pubchem.ncbi.nlm.nih.gov/compound/104289

      Helpful?

  2. What is the anisotropy refractive index and anisotropy electric permittivity for product 338680, 4´-Octyl-4-biphenylcarbonitrile?

    1 answer
    1. Literature values for the anisotropy refractive index and anisotropy electric permittivity of product 338680, 4´-Octyl-4-biphenylcarbonitrile are as follows:ne=1.673no=1.516in 27oCe||=12.8e^=6.65De=6.15in 39.4oC

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service