Skip to Content
Merck
All Photos(1)

Key Documents

338656

Sigma-Aldrich

4′-(Hexyloxy)-4-biphenylcarbonitrile

liquid crystal (nematic), 96%

Synonym(s):

4′-(Hexyloxy)-4-biphenyl-carbonitrile, 4′-(Hexyloxy)[1,1′-biphenyl]-4-carbonitrile, 4′-Hexyloxy-4-cyanobiphenyl, 4′-Hexyloxybiphenylyl-4-carbonitrile, 4-(4-Hexyloxyphenyl)benzonitrile, 4-(n-Hexyloxy)-4′-cyanobiphenyl, 4-Cyano-4′-(hexyloxy)biphenyl, 4-Cyano-4′-n -hexyloxybiphenyl, 4-Hexyloxy-4′-cyanobiphenyl, p -Hexyloxy-p ′-cyanobiphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€109.00

€109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€109.00

About This Item

Linear Formula:
CH3(CH2)5OC6H4C6H4CN
CAS Number:
Molecular Weight:
279.38
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€109.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

liquid crystal (nematic)

mp

56-58 °C (lit.)

transition temp

crystalline phase to nematic phase 56 °C
nematic phase to isotropic phase 76 °C

SMILES string

CCCCCCOc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C19H21NO/c1-2-3-4-5-14-21-19-12-10-18(11-13-19)17-8-6-16(15-20)7-9-17/h6-13H,2-5,14H2,1H3

InChI key

YUYXUPYNSOFWFV-UHFFFAOYSA-N

General description

Also known as 6OCBs,these liquid crystals (LCs) are responsive to small changes in temperature, shear, electric (or magnetic) fields, and the structure of solid surfaces with which they are in contact.[1] In LCs exhibiting smectic phases, such as these, the molecules show a degree of translational order and tend to align themselves in layers.[1]

Application

Used to synthesize pH responsive block copolymers,[1] binary mixtures.[2]

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

228.2 °F

Flash Point(C)

109 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

pH-responsive aqueous/LC interfaces using SGLCP-b-polyacrylic acid block copolymers.
Lee DY, et al.
Soft Matter, 6(9), 1964-1970 (2010)
Separation of nematic and smectic-A potential effects on solute ordering in a series of binary 6OCB-8OCB mixtures
Burnell EE, et al.
Canadian Journal of Chemistry, 89(7), 900-908 (2011)

Articles

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service