Skip to Content
Merck
All Photos(1)

Key Documents

332291

Sigma-Aldrich

3,4-Di-O-acetyl-6-deoxy-L-glucal

98%

Synonym(s):

Di-O-acetyl-L-rhamnal

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€95.20

€95.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€95.20

About This Item

Empirical Formula (Hill Notation):
C10H14O5
CAS Number:
Molecular Weight:
214.22
Beilstein:
84954
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

€95.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

optical activity

[α]/D 55°, c = 1 in chloroform

refractive index

n20/D 1.454 (lit.)

bp

68-69 °C/0.06 mmHg (lit.)

density

1.116 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H]1OC=C[C@H](OC(C)=O)[C@H]1OC(C)=O

InChI

1S/C10H14O5/c1-6-10(15-8(3)12)9(4-5-13-6)14-7(2)11/h4-6,9-10H,1-3H3/t6-,9-,10-/m0/s1

InChI key

NDEGMKQAZZBNBB-JUWDTYFHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service