Skip to Content
Merck
All Photos(2)

Key Documents

331708

Sigma-Aldrich

Dibromobis(triphenylphosphine)nickel(II)

99%

Synonym(s):

NiBr2(PPh3)2, bis(triphenylphosphine)nickel(II) bromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€109.00
50 G
€515.00

€109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€109.00
50 G
€515.00

About This Item

Linear Formula:
[(C6H5)3P]2NiBr2
CAS Number:
Molecular Weight:
743.07
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€109.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

reaction suitability

core: nickel
reagent type: catalyst

mp

219-223 °C (lit.)

SMILES string

Br[Ni]Br.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.2BrH.Ni/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2

InChI key

QEKXARSPUFVXIX-UHFFFAOYSA-L

Application

Catalysts for the cross-coupling reactions, C-X bond reduction, homocoupling of Csp2 halides, displacement of aryl halides, and oligomerization of dienes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho, K. M.; Lam, C. H.; Luh, T.-Y.
The Journal of Organic Chemistry, 54, 4474-4474 (1989)
Takagi, K.; Sakakibara, Y.
Chemistry Letters (Jpn), 1957-1957 (1989)
Kalinin, V. N.
Synthesis, 413-413 (1992)
Iyoda, M.; Kuwatani, Y.; Oda, M.
Journal of the American Chemical Society, 111, 3761-3761 (1989)

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service