Skip to Content
Merck

Skip To

213934

Bis(triphenylphosphine)dicarbonylnickel

solid

Synonym(s):

Bis(triphenylphosphine)nickel(0)dicarbonyl, Dicarbonylbis(triphenylphosphine)nickel(0), Nickel(0)bis(triphenylphosphine) dicarbonyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Check Cart for Availability
€ 118,00
€ 88,50

About This Item

Linear Formula:
[(C6H5)3P]2Ni(CO)2
CAS Number:
Molecular Weight:
639.28
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12161600
EC Number:
235-851-9
MDL number:

€ 88,50

List Price€ 118,00Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Bis(triphenylphosphine)dicarbonylnickel,

form

solid

Quality Segment

reaction suitability

core: nickel, reagent type: catalyst

mp

206-209 °C (lit.)

storage temp.

2-8°C

SMILES string

[Ni].[C-]#[O+].[C-]#[O+].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.2CO.Ni/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2*1-2;/h2*1-15H;;;

InChI key

SLFKPACCQUVAPG-UHFFFAOYSA-N

Application

Catalyst for:
  • Cyclotrimerization reactions
  • Cyclooligomerization reactions

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
237116794279331708
reaction suitability

core: nickel, reagent type: catalyst

reaction suitability

core: nickel, reagent type: catalyst

reaction suitability

core: nickel, reagent type: catalyst

reaction suitability

core: nickel, reagent type: catalyst

form

solid

form

solid

form

solid

form

-

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

mp

206-209 °C (lit.)

mp

250 °C (dec.) (lit.)

mp

195-200 °C

mp

219-223 °C (lit.)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

13007-90-4

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters