Skip to Content
Merck
All Photos(2)

Key Documents

329673

Sigma-Aldrich

3,4-(Methylenedioxy)phenylacetic acid

98%

Synonym(s):

1,3-Benzodioxole-5-acetic acid, Homopiperonylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€147.00

€147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€147.00

About This Item

Empirical Formula (Hill Notation):
C9H8O4
CAS Number:
Molecular Weight:
180.16
Beilstein:
177739
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€147.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

125-129 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)Cc1ccc2OCOc2c1

InChI

1S/C9H8O4/c10-9(11)4-6-1-2-7-8(3-6)13-5-12-7/h1-3H,4-5H2,(H,10,11)

InChI key

ODVLMCWNGKLROU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-(Methylenedioxy)phenylacetic acid (Homopiperonylic acid) is a 1, 3-benzodioxole derivative and its standard molar energy of combustion in oxygen was evaluated.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Agostinha R Matos et al.
Organic & biomolecular chemistry, 2(6), 908-914 (2004-03-10)
The standard (p(o)= 0.1 MPa) molar energies of combustion in oxygen, at T= 298.15 K, of four 1,3-benzodioxole derivatives (sesamol, piperonyl alcohol, piperonylic acid and homopiperonylic acid) were measured by static bomb calorimetry. The values of the standard molar enthalpies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service