Skip to Content
Merck
All Photos(1)

Key Documents

325198

Sigma-Aldrich

2,3,6-Trifluorobenzoic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€23.70

€23.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€23.70

About This Item

Linear Formula:
F3C6H2CO2H
CAS Number:
Molecular Weight:
176.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€23.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

130-131 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1c(F)ccc(F)c1F

InChI

1S/C7H3F3O2/c8-3-1-2-4(9)6(10)5(3)7(11)12/h1-2H,(H,11,12)

InChI key

MGUPHQGQNHDGNK-UHFFFAOYSA-N

General description

Raman and FTIR spectra of the 2,3,6-tri-fluorobenzoic acid was studied.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Mukherjee et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 77(4), 787-794 (2010-09-11)
Raman and FTIR spectra of the 2,3,6-tri-fluorobenzoic acid molecule have been recorded in the regions 50-4000cm⁻¹ and 400-4000cm⁻¹, respectively. Vibrational frequencies have been calculated by employing DFT method in dimeric form in optimum state. SQM force field has also been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service