Skip to Content
Merck
All Photos(1)

Key Documents

529184

Sigma-Aldrich

2,4,5-Trifluorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€37.90

€37.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€37.90

About This Item

Linear Formula:
F3C6H2CHO
CAS Number:
Molecular Weight:
160.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€37.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.482 (lit.)

bp

168 °C (lit.)

density

1.408 g/mL at 25 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

Fc1cc(F)c(C=O)cc1F

InChI

1S/C7H3F3O/c8-5-2-7(10)6(9)1-4(5)3-11/h1-3H

InChI key

CYIFJRXFYSUBFW-UHFFFAOYSA-N

Application

2,4,5-Trifluorobenzaldehyde may be used in the synthesis of:
  • bis(2,4,5-trifluorophenyl)methanone[1][2]
  • 8-phenyl-7-tosyl-1-(2,4,5-trifluorophenyl)octahydro-1H-pyrano[3,4-c] pyridine[3]
  • (E)-4-methoxy-N′-(2,4,5-trifluorobenzylidene)-benzohydrazide monohydrate[4]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(E)-4-Methoxy-N'-(2, 4, 5-trifluorobenzylidene) benzohydrazide monohydrate.
Maheswari R, et al.
IUCrJ, 1(5), x160846-x160846 (2016)
Zachary R Woydziak et al.
Synthesis, 46(2), 158-164 (2014-06-11)
Fluorinated fluorophores are valuable tools for studies of biological systems. However, amine-reactive single-isomer derivatives of these compounds are often very expensive. To provide an inexpensive alternative, we report a practical synthesis of 4-carboxy-Pennsylvania Green methyl ester. Derivatives of this hydrophobic
Aminol initiated Prins cyclization for the synthesis of octahydro-1H-pyrano [3, 4-c] pyridine and hexahydro-1H-furo [3, 4-c] pyrrole derivatives.
Tetrahedron Letters, 55(34), 4817-4821 (2014)
Zachary R Woydziak et al.
The Journal of organic chemistry, 77(1), 473-481 (2011-11-25)
Fluorination of fluorophores can substantially enhance their photostability and improve spectroscopic properties. To facilitate access to fluorinated fluorophores, bis(2,4,5-trifluorophenyl)methanone was synthesized by treatment of 2,4,5-trifluorobenzaldehyde with a Grignard reagent derived from 1-bromo-2,4,5-trifluorobenzene, followed by oxidation of the resulting benzyl alcohol.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service