Skip to Content
Merck
All Photos(2)

Key Documents

324345

Sigma-Aldrich

4-Nitro-1,8-naphthalic anhydride

95%

Synonym(s):

4-Nitronaphthalene-1,8-dicarboxylic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€223.00
25 G
€759.00

€223.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€223.00
25 G
€759.00

About This Item

Empirical Formula (Hill Notation):
C12H5NO5
CAS Number:
Molecular Weight:
243.17
Beilstein:
237909
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€223.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

mp

226-229 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc2C(=O)OC(=O)c3cccc1c23

InChI

1S/C12H5NO5/c14-11-7-3-1-2-6-9(13(16)17)5-4-8(10(6)7)12(15)18-11/h1-5H

InChI key

LKOZHLXUWUBRDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitro-1,8-naphthalic anhydride is a yellow crystalline nitrocompound widely used in the field of dye synthesis and cellular imaging.

Application

4-Nitro-1,8-naphthalic anhydride can be used:
  • As a precursor to synthesize N-phenyl-amino-1,8-naphthalimide based fluorescent chemosensor to detect nitro-antibiotics at ppb level.[1]
  • As a building block to synthesize shape memory polymers due to its ability to undergo Diels-Alder reaction
  • As a fluorochrome substrate for nitrogen reductase for noninvasive hypoxia imaging in cancer detection.[2]
  • As a precursor to synthesize amphiphilic naphthalimide dyes with good color brilliancy.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desislava Staneva et al.
Sensors (Basel, Switzerland), 20(12) (2020-06-25)
In this study, a novel 6-(allylamino)-2-(2-(dimethylamino)ethyl)-1H-benzo[de]isoquinoline-1,3(2H)-dione (NI3) was synthesized and characterized. Its copolymer with styrene was also obtained. The photophysical characteristics of NI3 were investigated in organic solvents and the results were compared with those of its structural analogue, 2-allyl-6-((2-(dimethylamino)ethyl)amino)-1H-benzo[de]isoquinoline-1,3(2H)-dione
Jin Zhou et al.
Chemistry, an Asian journal, 11(19), 2719-2724 (2016-02-09)
A lysosome-targeting fluorescent off-on probe has been developed by one-step synthesis for detecting lysosomal nitroreductase and hypoxia. The probe is constructed by incorporating morpholine (a lysosome-targeting unit) into 4-nitro-1,8-naphthalimide (as a fluorochrome and specific substrate for nitroreductase), and the detection
Synthesis of disperse dyes derived from 4-amino-N-decyl-1, 8-naphthalimide and their dyeing properties on polyester fabrics
Umar Salami Ameuru, et al.
Dyes and Pigments, 157, 190-197 (2018)
Discriminative fluorescent sensing of nitro-antibiotics at ppb level using N-phenyl-amino-1,8-naphthalimides chemosensors
Ananthu Shanmughan, et al.
Results in Chemistry, 4, 100546-100546 (2022)
Naphthalimides in fluorescent imaging of tumor hypoxia ? An up-to-date review
Rashmi Kumari, et al.
Bioorganic Chemistry, 88, 102979-102979 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service