Skip to Content
Merck
All Photos(1)

Key Documents

316938

Sigma-Aldrich

3-Methoxyphenylacetyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2COCl
CAS Number:
Molecular Weight:
184.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.5392 (lit.)

bp

120 °C/2 mmHg (lit.)

density

1.184 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

COc1cccc(CC(Cl)=O)c1

InChI

1S/C9H9ClO2/c1-12-8-4-2-3-7(5-8)6-9(10)11/h2-5H,6H2,1H3

InChI key

UZUYKYNVSJTWEH-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service