Skip to Content
Merck
All Photos(1)

Key Documents

293164

Sigma-Aldrich

Acetyl chloride-13C2

99 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€258.00
1 G
€913.00

€258.00


Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
€258.00
1 G
€913.00

About This Item

Linear Formula:
13CH313COCl
CAS Number:
Molecular Weight:
80.48
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

€258.00


Check Cart for Availability

Request more information

vapor density

2.7 (vs air)

isotopic purity

99 atom % 13C

Assay

99% (CP)

autoignition temp.

1353 °F

expl. lim.

19 %

refractive index

n20/D 1.389 (lit.)

bp

52 °C (lit.)

mp

-112 °C (lit.)

density

1.130 g/mL at 25 °C

mass shift

M+2

storage temp.

2-8°C

SMILES string

[13CH3][13C](Cl)=O

InChI

1S/C2H3ClO/c1-2(3)4/h1H3/i1+1,2+1

InChI key

WETWJCDKMRHUPV-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service