Skip to Content
Merck
All Photos(2)

Key Documents

309257

Sigma-Aldrich

Tin(II) bromide

Synonym(s):

Stannous bromide, Stannous dibromide, Tin(2+) dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€43.20

€43.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€43.20

About This Item

Linear Formula:
SnBr2
CAS Number:
Molecular Weight:
278.52
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

€43.20


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

concentration

55.4-59.4% (Br, silver nitrate titration)

density

5.12 g/mL at 25 °C (lit.)

SMILES string

Br[SnH2]Br

InChI

1S/2BrH.Sn/h2*1H;/q;;+2/p-2

InChI key

ZSUXOVNWDZTCFN-UHFFFAOYSA-L

Related Categories

Application


  • Green chemistry: Aqueous synthesis of methylammonium tin bromide from tin (II) oxide for sustainable semiconductor applications: This work involves the synthesis of methylammonium tin bromide (MASnBr3) from methylammonium bromide and tin(II) bromide for semiconductor applications (Y Harsono, D Onggo, 2024).

  • A zero-dimensional organic seesaw-shaped tin bromide with highly efficient strongly stokes-shifted deep-red emission: The study reports an organic tin bromide hybrid that exhibits deep-red emission, synthesized from tin(II) bromide and 1-butyl-1-methylpyrrolidinium solutions (C Zhou et al., 2018).

  • Synthesis, Structure, and Bonding Analysis of Tin (II) Dihalide and Cyclopentadienyltin (II) Halide (Alkyl)(amino) carbene Complexes: This research explores the coordination chemistry of tin(II) bromide carbene complexes and their reactivity with iron tetracarbonyl (C Muller et al., 2019).

  • Two modifications of tin (II) bromide: The article investigates the crystal growth of tin(II) bromide under different temperature conditions, resulting in two modifications of the compound (P Eckold et al., 2015).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Nazarenko et al.
Chemistry of materials : a publication of the American Chemical Society, 31(6), 2121-2129 (2019-04-02)
Hybrid organic-inorganic main-group metal halide compounds are the subject of intense research owing to their unique optoelectronic characteristics. In this work, we report the synthesis, structure, and electronic and optical properties of a family of hybrid tin (II) bromide compounds
Adam J Biacchi et al.
ACS nano, 12(10), 10045-10060 (2018-09-25)
Colloidal-based solution syntheses offer a scalable and cost-efficient means of producing 2D nanomaterials in high yield. While much progress has been made toward the controlled and tailorable synthesis of semiconductor nanocrystals in solution, it remains a substantial challenge to fully

Questions

  1. What is the Purity?

    1 answer
    1. The purity of this product is not not determined. However, the minimum allowable Bromine content in this compound is 55.4% by titration. This indicates a minimum purity of 97%. This is a calculated estimate based on the molecular weight of this material and does not take into account any moisture or impurities that may be present.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service