Skip to Content
Merck
All Photos(1)

Key Documents

215961

Sigma-Aldrich

Tin(IV) bromide

99%

Synonym(s):

Tetrabromostannane, Tetrabromotin, Tin tetrabromide, Stannic bromide

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€121.00

€121.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€121.00

About This Item

Linear Formula:
SnBr4
CAS Number:
Molecular Weight:
438.33
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€121.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

reaction suitability

core: tin
reagent type: catalyst

bp

202 °C (lit.)

mp

31 °C (lit.)

density

3.34 g/mL at 25 °C (lit.)

SMILES string

Br[Sn](Br)(Br)Br

InChI

1S/4BrH.Sn/h4*1H;/q;;;;+4/p-4

InChI key

LTSUHJWLSNQKIP-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongkun Yu et al.
The journal of physical chemistry. B, 123(23), 4958-4966 (2019-05-24)
The concept of eutectic molecular liquids (EMLs) was defined, and a strategy to form EMLs based on noncovalent interactions was proposed. We verified the formation and investigated the properties, interaction sites, and interaction energies of the obtained 16 EMLs. Moreover
Adam J Biacchi et al.
ACS nano, 12(10), 10045-10060 (2018-09-25)
Colloidal-based solution syntheses offer a scalable and cost-efficient means of producing 2D nanomaterials in high yield. While much progress has been made toward the controlled and tailorable synthesis of semiconductor nanocrystals in solution, it remains a substantial challenge to fully

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service