Skip to Content
Merck
All Photos(1)

Key Documents

303690

Sigma-Aldrich

Hexaammineruthenium(II) chloride

99.9% trace metals basis

Synonym(s):

Ruthenium(III) chloride hexaammoniate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€93.40
1 G
€279.00
5 G
€1,050.00

€93.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€93.40
1 G
€279.00
5 G
€1,050.00

About This Item

Linear Formula:
[Ru(NH3)6]Cl2
CAS Number:
Molecular Weight:
273.15
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid
solubility:
H2O: slightly soluble(lit.)

€93.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.9% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: ruthenium

impurities

≤1500.0 ppm Trace Metal Analysis

solubility

H2O: slightly soluble(lit.)

storage temp.

2-8°C

SMILES string

N.N.N.N.N.N.[Cl-].[Cl-].[Ru++]

InChI

1S/2ClH.6H3N.Ru/h2*1H;6*1H3;/q;;;;;;;;+2/p-2

InChI key

SHHNQLHKJVODTN-UHFFFAOYSA-L

General description

Hexaammineruthenium(II) chloride is a yellow crystalline coordination compound that is slightly soluble in water. It acts as a catalyst in a wide range of chemical reactions, including hydrogenation, isomerization, and oxidation reactions. It is also used as a redox probe in electroanalytical techniques.[1] [2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activated Carbon-Supported Ruthenium as a Catalyst for the Solvent- and Initiator-Free Aerobic Epoxidation of Limonene
Sara Madadi and Serge Kaliaguine
ACS sustainable chemistry & engineering, 9, 10557-10568 (2021)
Effect of Nanoporous Gold Thin Film Morphology on Electrochemical DNA Sensing
Pallavi Daggumati, et al.
Analytical Chemistry, 87, 8149-8156 (2015)
Giulia Selvolini et al.
Sensors (Basel, Switzerland), 19(14) (2019-07-25)
In this work, we report the development of a simple and sensitive sensor based on graphite screen-printed electrodes (GSPEs) modified by a nanocomposite film for dopamine (DA) detection. The sensor was realized by electrodepositing polyaniline (PANI) and gold nanoparticles (AuNPs)
Jiachao Yu et al.
Analytical chemistry, 86(16), 8129-8135 (2014-07-19)
Evaluating the kinetics of biological reaction occurring in confined nanospaces is of great significance in studying the molecular biological processes in vivo. Herein, we developed a nanochannel-based electrochemical reactor and a kinetic model to investigate the immunological reaction in confined

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service