Skip to Content
Merck
All Photos(2)

Key Documents

361356

Sigma-Aldrich

Pentamethylcyclopentadienylruthenium(III) chloride polymer

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€89.10

€89.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€89.10

About This Item

Linear Formula:
[C10H15Cl2Ru]n
CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

€89.10


Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

core: ruthenium
reaction type: solution phase peptide synthesis
reagent type: catalyst

SMILES string

Cl[Ru]Cl.Cl[Ru]Cl.C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]2[C](C)[C](C)[C](C)[C]2C

InChI

1S/2C10H15.4ClH.2Ru/c2*1-6-7(2)9(4)10(5)8(6)3;;;;;;/h2*1-5H3;4*1H;;/q;;;;;;2*+2/p-4

InChI key

VDXZXHGAMPUNED-UHFFFAOYSA-J

Application

Entry points into pentamethylcyclopentadienyl chemistry of ruthenium.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemische Berichte, 121, 1719-1719 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service