Skip to Content
Merck
All Photos(1)

Key Documents

302570

Sigma-Aldrich

N,N-Diethylpropionamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CON(C2H5)2
CAS Number:
Molecular Weight:
129.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

77 °C/12 mmHg (lit.)

density

0.897 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)C(=O)CC

InChI

1S/C7H15NO/c1-4-7(9)8(5-2)6-3/h4-6H2,1-3H3

InChI key

YKOQQFDCCBKROY-UHFFFAOYSA-N

General description

Gas phase ion/molecule reaction between dialkoxyborinium cation and N,N-diethylpropionamide was studied.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Zheng et al.
Journal of the American Society for Mass Spectrometry, 12(8), 948-955 (2001-08-17)
We report gas phase ion/molecule reactions between dialkoxyborinium cations (RO-B+-OR) and small organic amides, such as N,N-dimethylformamide and N,N-diethylpropionamide. Besides direct addition at boron, the results show efficient SN2 methyl transfer from the borinium ion to the amide. Isotopic labeling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service