Skip to Content
Merck
All Photos(2)

Key Documents

347159

Sigma-Aldrich

N,N-Dimethylisobutyramide

99%

Synonym(s):

N,N,2-Trimethylpropionamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€30.70

€30.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€30.70

About This Item

Linear Formula:
(CH3)2CHCON(CH3)2
CAS Number:
Molecular Weight:
115.17
Beilstein:
1742178
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€30.70


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

175-176 °C/744 mmHg (lit.)

density

0.892 g/mL at 25 °C (lit.)

functional group

amide

SMILES string

CC(C)C(=O)N(C)C

InChI

1S/C6H13NO/c1-5(2)6(8)7(3)4/h5H,1-4H3

InChI key

GXMIHVHJTLPVKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylisobutyramide is an N,N-disubstituted amide. It can be prepared by acylation of secondary amines with acyl chloride.[1]

Application

N,N-Dimethylisobutyramide may be used in the preparation of 1-chloro-N,N,2-trimethylpropenylamine.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

α-Chloro Enamines, Reactive Intermediates for Synthesis: 1-Chloro-N, N, 2-Trimethylpropenylamine.
Haveaux B, et al.
Organic Syntheses, 26-26 (1979)
Infrared spectroscopic studies of N, N-disubstituted amides as models for the peptide bond in hydrogen bonded interactions with water molecules.
Henson DB and Swenson CA.
The Journal of Physical Chemistry, 77(20), 2401-2406 (1973)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service