Skip to Content
Merck
All Photos(2)

Key Documents

301817

Sigma-Aldrich

5,6-Dimethyl-1,10-phenanthroline

97%

Synonym(s):

5,6-Dimethyl-o-phenanthroline

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€239.00
1 G
€300.00

€239.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€239.00
1 G
€300.00

About This Item

Empirical Formula (Hill Notation):
C14H12N2
CAS Number:
Molecular Weight:
208.26
Beilstein:
160918
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€239.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

266-269 °C (lit.)

SMILES string

Cc1c(C)c2cccnc2c3ncccc13

InChI

1S/C14H12N2/c1-9-10(2)12-6-4-8-16-14(12)13-11(9)5-3-7-15-13/h3-8H,1-2H3

InChI key

BRPQDJPJBCQFSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5,6-Dimethyl-1,10-phenanthroline was employed as ligand in the preparation of:
  • novel tetrahedral copper(I) mixed-ligand complexes[1]
  • mixed ligand Ru(II) complexes[2]
  • 5,6-bis(bromomethyl)-1,10-phenanthroline via bromination with N-bromosuccinimide[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Gandin et al.
Journal of medicinal chemistry, 56(18), 7416-7430 (2013-08-24)
Novel tetrahedral copper(I) mixed-ligand complexes of the type [Cu(X)(N(∩)N)(PCN)], 3-10, where X = Cl or Br, N(∩)N = 2,2'-bipyridine (bipy), 1,10-phenanthroline (phen), 5,6-dimethyl-1,10-phenanthroline (dmp), and dipyrido-[3,2-d:2',3'-f]-quinoxaline (dpq), and PCN = tris-(2-cyanoethyl)phosphine, have been synthetized and characterized by NMR, ESI-MS, and
Mixed ligand ruthenium (II) complexes of 5, 6-dimethyl-1, 10-phenanthroline: the role of ligand hydrophobicity on DNA binding of the complexes.
Uma MP, et al.
Inorgorganica Chimica Acta, 359(14), 4601-4612 (2006)
Self-assembled fullerene-derivative monolayers on a gold substrate using phenanthroline-Au interactions.
Dominguez O, et al.
Langmuir, 14(4), 821-824 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service