Skip to Content
Merck
All Photos(1)

Key Documents

301779

Sigma-Aldrich

5-Chloro-1,10-phenanthroline

98%

Synonym(s):

5-Chloro-1,10-diazaphenanthrene, 5-Chloro-o-phenanthroline

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€92.20

€92.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€92.20

About This Item

Empirical Formula (Hill Notation):
C12H7ClN2
CAS Number:
Molecular Weight:
214.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€92.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

124-126 °C (lit.)

functional group

chloro

SMILES string

Clc1cc2cccnc2c3ncccc13

InChI

1S/C12H7ClN2/c13-10-7-8-3-1-5-14-11(8)12-9(10)4-2-6-15-12/h1-7H

InChI key

XDUUQOQFSWSZSM-UHFFFAOYSA-N

Application

5-Chloro-1,10-phenanthroline was used in preparation of:
  • 1,10-phenanthrolinium cation, new building block for the design of ionic liquid crystals[1]
  • platinum(II)-based metallointercalators[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Cardinaels et al.
Langmuir : the ACS journal of surfaces and colloids, 27(5), 2036-2043 (2011-01-22)
The 1,10-phenanthrolinium cation is introduced as a new building block for the design of ionic liquid crystals. 1,10-Phenanthroline, 5-methyl-1,10-phenanthroline, 5-chloro-1,10-phenanthroline, and 4,7-diphenyl-1,10-phenanthroline were quaternized by reaction with 1,3-dibromopropane or 1,2-dibromoethane. The resulting cations were combined with dodecyl sulfate or dioctyl
Sharon Kemp et al.
Journal of inorganic biochemistry, 101(7), 1049-1058 (2007-06-05)
Fifteen platinum(II)-based metallointercalators have been synthesised that utilise substituted 1,10-phenanthroline (phen) ligands, including 5-chloro-1,10-phenanthroline (5-Cl-phen), 5-methyl-1,10-phenanthroline (5-CH3-phen), 5-amino-1,10-phenanthroline (5-NH2-phen), 5-nitro-1,10-phenanthroline (5-NO2-phen) and dipyrido[3,2-d:2',3'-f]quinoxaline (dpq), and achiral ethylenediamine (en) and the chiral ancillary ligands 1S,2S-diaminocyclohexane (S,S-dach) and 1R,2R-diaminocyclohexane (R,R-dach). Their cytotoxicity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service