Skip to Content
Merck
All Photos(1)

Key Documents

301426

Sigma-Aldrich

Hydrobenzoin

Synonym(s):

1,2-Diphenyl-1,2-ethanediol

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€44.60

€44.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€44.60

About This Item

Linear Formula:
C6H5CH(OH)CH(OH)C6H5
CAS Number:
Molecular Weight:
214.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€44.60


Check Cart for Availability

Request a Bulk Order

SMILES string

OC(C(O)c1ccccc1)c2ccccc2

InChI

1S/C14H14O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-16H

InChI key

IHPDTPWNFBQHEB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Toselli et al.
Drug metabolism and disposition: the biological fate of chemicals, 45(8), 966-973 (2017-06-11)
Oxetanyl building blocks are increasingly used in drug discovery because of the improved drug-like properties they confer on drug candidates, yet little is currently known about their biotransformation. A series of oxetane-containing analogs was studied and we provide the first
Randy M Wadkins et al.
Journal of medicinal chemistry, 48(8), 2906-2915 (2005-04-15)
Carboxylesterases (CE) are ubiquitous enzymes responsible for the metabolism of xenobiotics. Because the structural and amino acid homology among esterases of different classes, the identification of selective inhibitors of these proteins has proved problematic. Using Telik's target-related affinity profiling (TRAP)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service