HPLC Analysis of Hydrobenzoin Enantiomers on Astec® CYCLOBOND I 2000
![HPLC Analysis of Hydrobenzoin Enantiomers on Astec® CYCLOBOND I 2000 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/233/627/36ce3ff2-ab6e-40f9-9022-f0e9b9140630/800/36ce3ff2-ab6e-40f9-9022-f0e9b9140630.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] acetonitrile; (80:20, A:B)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null