Skip to Content
Merck
All Photos(1)

Key Documents

245003

Sigma-Aldrich

Dirhenium decacarbonyl

98%

Synonym(s):

Rhenium carbonyl

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€464.00

€464.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€464.00

About This Item

Linear Formula:
Re2(CO)10
CAS Number:
Molecular Weight:
652.52
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€464.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder or crystals

reaction suitability

core: rhenium
reagent type: catalyst

mp

170 °C (dec.) (lit.)

SMILES string

[ReH].[ReH].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/10CO.2Re/c10*1-2;;

InChI key

ZIZHEHXAMPQGEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dirhenium decacarbonyl (Re2(CO)10) is a transition metal carbonyl widely used as a precursor in the chemical vapor deposition (CVD) of metals and the fabrication of composite materials. The endothermic nature of the decomposition of Re2(CO)10 allows for uniform deposition of metallic or alloy coatings on the substrate.

Application

Dirhenium decacarbonyl can be used as a precursor to synthesize:
  • Re nanoparticles via pulsed-laser decomposition.
  • Amide functionalized Re(I) based dinuclear metallacycles that can selectively inhibit certain cancer cells.
  • Twisted ultrathin rhenium disulfide nanoribbons encapsulated in single-walled carbon nanotubes.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petros M Sakkas et al.
Ultrasonics sonochemistry, 69, 105250-105250 (2020-07-16)
Rhenium oxide nanoparticles have been prepared using ultrasonication at 20 kHz. Samples characterization was committed via SEM-EDX, TEM, XRD, and Raman spectroscopy. Various experimental parameters were examined, including precursor/substrate amounts, ultrasonication intensity, and type of solvent used. Insights to the agglomeration
Azimkhan Kozhakhmetov et al.
Advanced materials (Deerfield Beach, Fla.), 32(50), e2005159-e2005159 (2020-11-11)
Reliable, controlled doping of 2D transition metal dichalcogenides will enable the realization of next-generation electronic, logic-memory, and magnetic devices based on these materials. However, to date, accurate control over dopant concentration and scalability of the process remains a challenge. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service