Skip to Content
Merck
All Photos(2)

Documents

295531

Sigma-Aldrich

Methylamine

anhydrous, ≥98%

Synonym(s):

Monomethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH2
CAS Number:
Molecular Weight:
31.06
Beilstein:
741851
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

vapor density

1.08 (20 °C, vs air)

vapor pressure

27 psi ( 20 °C)

Assay

≥98%

autoignition temp.

806 °F

expl. lim.

4.9-20.8 %

bp

-6.3 °C (lit.)

mp

-93 °C (lit.)

density

0.70 g/mL at 20 °C (lit.)

SMILES string

CN

InChI

1S/CH5N/c1-2/h2H2,1H3

InChI key

BAVYZALUXZFZLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methylamine may be used to introduce N-methyl groups into the polymer backbone during the synthesis of Si-methyl- and N-methyl-containing polyborosilazane ([B(C2H4SiCH3NCH3)3]n) from tris(dichloromethylsilylethyl)borane.

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Compr. Gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron-modified polysilazane as a novel single-source precursor for SiBCN ceramic fibers: synthesis, melt-spinning, curing and ceramic conversion.
Bernard S, et al.
Journal of Materials Chemistry, 15(2), 289-299 (2005)
Abdelaziz Al Ouahabi et al.
Nature communications, 8(1), 967-967 (2017-10-19)
In the context of data storage miniaturization, it was recently shown that digital information can be stored in the monomer sequences of non-natural macromolecules. However, the sequencing of such digital polymers is currently limited to short chains. Here, we report
Sebastian Strauss et al.
Nature methods, 15(9), 685-688 (2018-08-22)
Although current implementations of super-resolution microscopy are technically approaching true molecular-scale resolution, this has not translated to imaging of biological specimens, because of the large size of conventional affinity reagents. Here we introduce slow off-rate modified aptamers (SOMAmers) as small
Xin Yu Chin et al.
Nature communications, 6, 7383-7383 (2015-06-26)
Despite the widespread use of solution-processable hybrid organic-inorganic perovskites in photovoltaic and light-emitting applications, determination of their intrinsic charge transport parameters has been elusive due to the variability of film preparation and history-dependent device performance. Here we show that screening
Katja Wosikowski et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 9(5), 1917-1926 (2003-05-10)
To avoid systemic toxicity of the cytotoxic drug methotrexate (MTX) and to improve tumor selectivity, MTX was bound to human serum albumin (HSA) as a drug carrier. To understand more about the mechanism of action of MTX conjugated to HSA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service