Skip to Content
Merck
All Photos(1)

Key Documents

295361

Sigma-Aldrich

Hexafluoroethane

≥98%

Synonym(s):

CFC-116, Perfluoroethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF3
CAS Number:
Molecular Weight:
138.01
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.7 (vs air)

vapor pressure

430 psi ( 21 °C)

Assay

≥98%

bp

−78 °C (lit.)

mp

−100 °C (lit.)

application(s)

PFAS testing

functional group

fluoro

SMILES string

FC(F)(F)C(F)(F)F

InChI

1S/C2F6/c3-1(4,5)2(6,7)8

InChI key

WMIYKQLTONQJES-UHFFFAOYSA-N

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Management of bilateral acute hydrops secondary to keratoglobus with perfluoroethane (C2F6) pneumodescemetopexy.
Christine Kiire et al.
Clinical & experimental ophthalmology, 37(9), 892-894 (2010-01-23)
Harvey Lincoff et al.
Retina (Philadelphia, Pa.), 28(1), 56-59 (2008-01-11)
To test the validity of the geometric conclusion that 40 degrees gaze down is optimal for pneumatic displacement of a subretinal hemorrhage (SRH) in the macula. Nine consecutive patients with SRH in the macula had an intravitreal injection of perfluorocarbon
[Long-lasting expanding gases for the treatment of vitreo-retinal diseases].
Akito Hirakata
Nippon Ganka Gakkai zasshi, 112(1), 3-6 (2008-02-05)
Rubina Rahman et al.
The British journal of ophthalmology, 96(2), 185-188 (2011-05-19)
To compare the outcomes of non-posturing macular-hole surgery using sulfur hexafluoride (SF(6)) gas versus perfluoroethane (C(2)F(6)) for idiopathic macular hole repair. Design Interventional, comparative cohort study. 39 eyes of 38 patients undergoing macular-hole surgery with SF(6) were compared with another
Aurore Muselier et al.
American journal of ophthalmology, 150(3), 387-391 (2010-07-10)
To compare the functional and the anatomic outcomes of a combined surgery and consecutive surgery for macular hole and cataract extraction. Multicenter, retrospective, comparative case series. One hundred twenty patients (120 eyes) with an idiopathic macular hole and cataract were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service