Skip to Content
Merck
All Photos(1)

Key Documents

290556

Sigma-Aldrich

3-(Trifluoromethyl)benzyl bromide

98%

Synonym(s):

α′-Bromo-α,α,α-trifluoro-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2Br
CAS Number:
Molecular Weight:
239.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

refractive index

n20/D 1.492 (lit.)

bp

69 °C/4 mmHg (lit.)

density

1.565 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

FC(F)(F)c1cccc(CBr)c1

InChI

1S/C8H6BrF3/c9-5-6-2-1-3-7(4-6)8(10,11)12/h1-4H,5H2

InChI key

MYYYZNVAUZVXBO-UHFFFAOYSA-N

Application

3-(Trifluoromethyl)benzyl bromide has been used in the preparation of flocoumafen[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthetic approaches and biological activities of 4-hydroxycoumarin derivatives.
Jung J-C and Park O-S.
Molecules (Basel), 14(11), 4790-4803 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service