Skip to Content
Merck
All Photos(1)

Key Documents

277142

Sigma-Aldrich

1-Bromo-2-methyl-1-propene

98%

Synonym(s):

Isocrotyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€92.20
50 G
€333.00

€92.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€92.20
50 G
€333.00

About This Item

Linear Formula:
(CH3)2C=CHBr
CAS Number:
Molecular Weight:
135.00
Beilstein:
1733844
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€92.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

92 °C (lit.)

density

1.318 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

storage temp.

2-8°C

SMILES string

C\C(C)=C\Br

InChI

1S/C4H7Br/c1-4(2)3-5/h3H,1-2H3

InChI key

DEFNUDNHTUZJAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Bromo-2-methyl-1-propene has been used in:
  • preparation of esters of 2,4-dienoic acids via palladium catalyzed reaction with methyl acrylate in the presence of triethylamine[1]
  • total synthesis of ocular age pigment, A2-E[2]
  • synthesis of 3-nor-geraniol [2H2][3]

Other Notes

May darken in storage

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed conjugated diene synthesis from vinylic halides and olefinic compounds.
Dieck HA and Heck RF.
The Journal of Organic Chemistry, 40(8), 1083-1090 (1975)
Biosynthesis of iridoid monoterpenes in insects: Defensive secretions from larvae of leaf beetles (coleoptera: chrysomelidae).
Veith M, et al.
Tetrahedron, 50(23), 6859-6874 (1994)
Total synthesis of the ocular age pigment A2-E: a convergent pathway.
Ren R X-F, et al.
Journal of the American Chemical Society, 119(15), 3619-3620 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service