Skip to Content
Merck
All Photos(1)

Key Documents

273155

Sigma-Aldrich

Acetone azine

98%

Synonym(s):

1,1,4,4-Tetramethyl-2,3-diazabuta-1,3-diene, 2-Propanone 2-(1-methylethylidene)hydrazone, Acetazine, Acetone ketazine, Dimethyl ketazine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€153.00

€153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€153.00

About This Item

Linear Formula:
(CH3)2C=NN=C(CH3)2
CAS Number:
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.02

€153.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

133 °C/763 mmHg (lit.)

mp

−125 °C (lit.)

density

0.842 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

C\C(C)=N/N=C(/C)C

InChI

1S/C6H12N2/c1-5(2)7-8-6(3)4/h1-4H3

InChI key

PFLUPZGCTVGDLV-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of methyl-substituted titanocene?bis(trimethylsilyl)acetylene complexes with acetone azine: crystal structures of (η5:η1-C5HMe3CH2CMe2NH)2Ti and (C5Me5)2Ti(NCMe2)
Rep M, et al.
Journal of Organometallic Chemistry, 597(1), 146-156 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service