Skip to Content
Merck
All Photos(1)

Key Documents

269042

Sigma-Aldrich

2,2,2-Trifluoroethylamine

99.5%

Synonym(s):

2-Amino-1,1,1-trifluoroethane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€53.10
5 G
€157.00

€53.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€53.10
5 G
€157.00

About This Item

Linear Formula:
CF3CH2NH2
CAS Number:
Molecular Weight:
99.06
Beilstein:
1733204
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€53.10


Check Cart for Availability

Request a Bulk Order

vapor pressure

~7.6 psi ( 20 °C)

Assay

99.5%

form

liquid

refractive index

n20/D 1.301 (lit.)

bp

36-37 °C (lit.)

density

1.262 g/mL at 20 °C (lit.)

functional group

amine
fluoro

storage temp.

2-8°C

SMILES string

NCC(F)(F)F

InChI

1S/C2H4F3N/c3-2(4,5)1-6/h1,6H2

InChI key

KIPSRYDSZQRPEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,2-Trifluoroethylamine, a volatile organic compound, was tested using corona discharge ion mobility spectrometry with orthogonal acceleration time of flight mass spectrometry (CD IMS-oaTOF)[1].

Application

2,2,2-Trifluoroethylamine was used in the determination of ibuprofen (2-(p-isobutylphenyl)propionic acid), a non-steroid anti-inflammatory drug (NSAID) residue in surface and wastewater samples[2].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

3.2 °F - closed cup

Flash Point(C)

-16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana María Casas Ferreira et al.
Journal of chromatography. A, 1218(30), 4856-4862 (2011-03-16)
A method using headspace generation and in situ derivatization with water soluble EDC (1-[3-(dimethylamino)propyl]-3-ethylcarbodiimide) and TFEA (2,2,2-trifluoroethylamine) has been optimized for the determination of ibuprofen (2-(p-isobutylphenyl)propionic acid), one of the most common non-steroid anti-inflammatory drug (NSAIDs) residues in surface and
Martin Sabo et al.
Analytical chemistry, 84(12), 5327-5334 (2012-05-19)
We demonstrate the application of corona discharge ion mobility spectrometry with orthogonal acceleration time of flight mass spectrometry (CD IMS-oaTOF) for volatile organic compounds (VOCs) monitoring. Two-dimensional (2D) IMS-oaTOF spectra of VOCs were recorded in nearly real time. The corona
The many roles for fluorine in medicinal chemistry.
William K Hagmann
Journal of medicinal chemistry, 51(15), 4359-4369 (2008-06-24)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service