Skip to Content
Merck
All Photos(1)

Key Documents

26580

Sigma-Aldrich

5-Chlorovaleric acid

≥98.0% (GC)

Synonym(s):

5-Chloropentanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€110.00

€110.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€110.00

About This Item

Linear Formula:
Cl(CH2)4COOH
CAS Number:
Molecular Weight:
136.58
Beilstein:
1745183
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€110.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.454 (lit.)
n20/D 1.454

mp

18 °C (lit.)

density

1.170 g/mL at 20 °C (lit.)

functional group

carboxylic acid
chloro

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCl

InChI

1S/C5H9ClO2/c6-4-2-1-3-5(7)8/h1-4H2,(H,7,8)

InChI key

YSXDKDWNIPOSMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Chlorovaleric acid was used to derivatize L-hydroxyproline for the synthesis of haptens HP1 and HP2. These haptens were used in the preparation of antibodies against L-hydroxyproline[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li L Wang et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(1), 9-15 (2012-10-04)
China's government has prohibited the addition of simply hydrolyzed animal protein from solid leather waste into milk. The objective of this study was to produce a monoclonal antibody against L-hydroxyproline, a special amino acid in hydrolyzed animal protein. L-hydroxyproline was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service