Skip to Content
Merck
All Photos(1)

Key Documents

C73001

Sigma-Aldrich

5-Chlorovaleronitrile

98%

Synonym(s):

5-Chloropentanenitrile

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€48.70

€48.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€48.70

About This Item

Linear Formula:
Cl(CH2)4CN
CAS Number:
Molecular Weight:
117.58
Beilstein:
1742079
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€48.70


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.446 (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

ClCCCCC#N

InChI

1S/C5H8ClN/c6-4-2-1-3-5-7/h1-4H2

InChI key

JSAWFGSXRPCFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service