Skip to Content
Merck
All Photos(2)

Key Documents

26475

Sigma-Aldrich

O-(p-Tolyl) chlorothionoformate

≥97.0%

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€157.00

€157.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
10 ML
€157.00

About This Item

Linear Formula:
ClC(S)OC6H4CH3
CAS Number:
Molecular Weight:
186.66
Beilstein:
1448986
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€157.00


Estimated to ship on23 May 2025


Request a Bulk Order

Assay

≥97.0%

refractive index

n20/D 1.573 (lit.)
n20/D 1.573

bp

245 °C (lit.)
50 °C/0.2 mmHg (lit.)

density

1.225 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

Cc1ccc(OC(Cl)=S)cc1

InChI

1S/C8H7ClOS/c1-6-2-4-7(5-3-6)10-8(9)11/h2-5H,1H3

InChI key

UNCAXIZUVRKBMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

O-(p-Tolyl) chlorothionoformate has been used in the synthesis of:
  • α-L-2′-deoxythreofuranosyl nucleoside analogs[1]
  • alkenes from hindered alcohols[2][3][4]

Other Notes

Synthesis of alkenes from hindered alcohols[2][3][4]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Gerlach et al.
Helvetica Chimica Acta, 55, 2277-2277 (1972)
A.P. Neary et al.
Journal of the Chemical Society. Chemical Communications, 1090-1090 (1989)
Kiran S Toti et al.
European journal of medicinal chemistry, 46(9), 3704-3713 (2011-06-15)
The synthesis of a series of α-L-2'-deoxythreofuranosyl nucleosides featuring the nucleobases A, T, C and U is described in seven steps from 1,2-O-isopropyledene-α-L-threose, involving a Vorbrüggen coupling and a Barton-McCombie deoxygenation protocol as the key steps. All analogues, including a
H. Gerlach et al.
Journal of the Chemical Society. Chemical Communications, 1215-1215 (1972)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service