Skip to Content
Merck
All Photos(1)

Key Documents

259659

Sigma-Aldrich

Methyl 2-methoxybenzoate

99%

Synonym(s):

Methyl o-anisate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€35.10

€35.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€35.10

About This Item

Linear Formula:
CH3OC6H4CO2CH3
CAS Number:
Molecular Weight:
166.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€35.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.534 (lit.)

bp

248 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccccc1OC

InChI

1S/C9H10O3/c1-11-8-6-4-3-5-7(8)9(10)12-2/h3-6H,1-2H3

InChI key

PFYHAAAQPNMZHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

FeCl2 catalyzed cross-coupling reaction between methyl 2-methoxybenzoate and diphenylmethane under various reaction conditions has been investigated[1].

Application

Methyl 2-methoxybenzoate has been used in the synthesis of (+/−)-vibralactone[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Zhou Li et al.
Angewandte Chemie (International ed. in English), 48(21), 3817-3820 (2009-04-18)
Hooking up: FeCl(2) catalyzes the efficient cross dehydrogenative arylation of substrates having benzylic C-H bonds (see scheme). High regioselectivity was observed during the cross-coupling between compounds containing aromatic C(sp(2))-H bonds and benzylic C(sp(3))-H bonds. This process is proposed to proceed
Quan Zhou et al.
Organic letters, 10(7), 1401-1404 (2008-03-04)
Reductive alkylation of methyl 2-methoxybenzoate with prenyl bromide and hydrolysis afforded methyl 6-oxo-1-prenyl-2-cyclohexenecarboxylate. Reduction of the ketone, hydrolysis, iodolactonization, ozonolysis, and intramolecular aldol reaction provided a spiro lactone cyclopentenal. Retro-iodolactonization with activated Zn, formation of the beta-lactone, and reduction of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service