Skip to Content
Merck
All Photos(1)

Key Documents

Z183393

Foil/poly bag

size 4 1/2 in. × 8 in.

Synonym(s):

Polyethylene Bag

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€68.70

€68.70


Check Cart for Availability


Select a Size

Change View
1 PKG
€68.70

About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

€68.70


Check Cart for Availability

packaging

pkg of 50 ea

size

4 1/2 in. × 8 in.

thickness

5 mil

Looking for similar products? Visit Product Comparison Guide

General description

Heat sealable. Useful for storing air, light, or moisture-sensitive reagents, odoriferous products, and for temporarily containing leaking bottles.

Recommended heat seal conditions: 400°F (204°C), 40 psi, 1.5 sec.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How many units come in the package?

    1 answer
    1. As listed in the 'PROPERTIES' section of the Product Detail Page, this product is supplied as a pack of 50 bags.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service