Skip to Content
Merck
All Photos(3)

Key Documents

251887

Sigma-Aldrich

2,4-Dimethoxyphenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€80.50

€80.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€80.50

About This Item

Linear Formula:
(CH3O)2C6H3NCO
CAS Number:
Molecular Weight:
179.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€80.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

bp

140 °C/11 mmHg (lit.)

mp

30 °C (lit.)

functional group

isocyanate

SMILES string

COc1ccc(N=C=O)c(OC)c1

InChI

1S/C9H9NO3/c1-12-7-3-4-8(10-6-11)9(5-7)13-2/h3-5H,1-2H3

InChI key

WRAZHLRMDRTLOZ-UHFFFAOYSA-N

Application

2,4-Dimethoxyphenyl isocyanate has been used in the preparation of pyrrolo[3,2-d]pyrimidines[1].

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frederik J R Rombouts et al.
Journal of combinatorial chemistry, 7(4), 589-598 (2005-07-12)
The first solid-phase methodology for the preparation of pyrrolo[3,2-d]pyrimidines is presented. Merrifield resin bearing a cysteamine "traceless" linker was treated with 4-oxo-N-(PhF)proline benzyl ester (10; PhF = 9-(9-phenylfluorenyl)) to provide resin-bound aminopyrrole 20, which was treated with ethyl, phenyl, 4-phenoxyphenyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service