Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

248703

Sigma-Aldrich

4-(4-Nitrophenylsulfonyl)aniline

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SO2C6H4NH2
CAS Number:
Molecular Weight:
278.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

96%

mp

169-171 °C (lit.)

SMILES string

Nc1ccc(cc1)S(=O)(=O)c2ccc(cc2)[N+]([O-])=O

InChI

1S/C12H10N2O4S/c13-9-1-5-11(6-2-9)19(17,18)12-7-3-10(4-8-12)14(15)16/h1-8H,13H2

InChI key

DMZVYFFBWHBWMO-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Uetrecht et al.
The Journal of pharmacology and experimental therapeutics, 245(1), 274-279 (1988-04-01)
Dapsone is an effective anti-inflammatory agent in conditions in which inflammation is mediated by neutrophils. Dapsone also has been associated with agranulocytosis. We found that neutrophils, which had been activated by a phorbol ester or opsonized zymosan, oxidized dapsone to
Stefano Pegoraro et al.
Nature communications, 8, 14193-14193 (2017-02-01)
Severe malaria is a life-threatening complication of an infection with the protozoan parasite Plasmodium falciparum, which requires immediate treatment. Safety and efficacy concerns with currently used drugs accentuate the need for new chemotherapeutic options against severe malaria. Here we describe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service