Merck
  • SC83288 is a clinical development candidate for the treatment of severe malaria.
SC83288 is a clinical development candidate for the treatment of severe malaria.
Nature communications (2017-02-01)
Stefano Pegoraro, Maëlle Duffey, Thomas D Otto, Yulin Wang, Roman Rösemann, Roland Baumgartner, Stefanie K Fehler, Leonardo Lucantoni, Vicky M Avery, Alicia Moreno-Sabater, Dominique Mazier, Henri J Vial, Stefan Strobl, Cecilia P Sanchez, Michael Lanzer
ABSTRACT
Severe malaria is a life-threatening complication of an infection with the protozoan parasite Plasmodium falciparum, which requires immediate treatment. Safety and efficacy concerns with currently used drugs accentuate the need for new chemotherapeutic options against severe malaria. Here we describe a medicinal chemistry program starting from amicarbalide that led to two compounds with optimized pharmacological and antiparasitic properties. SC81458 and the clinical development candidate, SC83288, are fast-acting compounds that can cure a P. falciparum infection in a humanized NOD/SCID mouse model system. Detailed preclinical pharmacokinetic and toxicological studies reveal no observable drawbacks. Ultra-deep sequencing of resistant parasites identifies the sarco/endoplasmic reticulum Ca
Sigma-Aldrich
N6-Cyclopentyladenosine, solid