Skip to Content
Merck
All Photos(1)

Key Documents

242659

Sigma-Aldrich

2-Bromothiophenol

97%

Synonym(s):

2-Bromobenzenethiol

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€107.00
50 G
€365.00

€107.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€107.00
50 G
€365.00

About This Item

Linear Formula:
BrC6H4SH
CAS Number:
Molecular Weight:
189.07
Beilstein:
2039769
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€107.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.635 (lit.)

bp

128-130 °C/25 mmHg (lit.)

density

1.573 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Sc1ccccc1Br

InChI

1S/C6H5BrS/c7-5-3-1-2-4-6(5)8/h1-4,8H

InChI key

YUQUNWNSQDULTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Bromothiophenol (2-Bromobenzenethiol) reacts with 4-(4-bromo-3-nitro-phenyl)morpholine to form nitro-sulphide[1].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenothiazynes: preparation of 2-and 3-tertiary aminophenothiazines.
Jones DH.
Journal of the Chemical Society, 132-137 (1971)
Javier Campanini-Salinas et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-22)
A rapid emergence of resistant bacteria is occurring worldwide, endangering the efficacy of antibiotics and reducing the therapeutic arsenal available for treatment of infectious diseases. In the present study, we developed a new class of compounds with antibacterial activity obtained
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service