Skip to Content
Merck

Key Documents

Skip To

242578

3-Chloroiodobenzene

98%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Check Cart for Availability
€ 43,20
€ 32,40

About This Item

Linear Formula:
ClC6H4I
CAS Number:
Molecular Weight:
238.45
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
210-920-6
MDL number:
Assay:
98%
Form:
liquid

€ 32,40

List Price€ 43,20Save 25%
Web-Only Promotion

Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

98%

form

liquid

refractive index

n20/D 1.631 (lit.)

bp

103-104 °C/15 mmHg (lit.)

density

1.926 g/mL at 25 °C (lit.)

functional group

chloro, iodo

SMILES string

Clc1cccc(I)c1

InChI

1S/C6H4ClI/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

JMLWXCJXOYDXRN-UHFFFAOYSA-N

General description

The multiple-quantum proton NMR spectra of 3-chloroiodobenzene was studied.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
192546338737337285
description

98%

description

99%

description

97%

description

97%

assay

98%

assay

99%

assay

97%

assay

97%

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

bp

103-104 °C/15 mmHg (lit.)

bp

-

bp

153-154 °C/13 mmHg (lit.)

bp

69 °C/43 mmHg (lit.)

density

1.926 g/mL at 25 °C (lit.)

density

-

density

-

density

1.46 g/mL at 25 °C (lit.)

refractive index

n20/D 1.631 (lit.)

refractive index

-

refractive index

-

refractive index

n20/D 1.465 (lit.)

form

liquid

form

solid

form

solid

form

solid


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

215.6 °F - closed cup

Flash Point (°C)

102.00 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters