Select a Size
About This Item
Recommended Products
Assay
99%
form
solid
mp
56-58 °C (lit.)
functional group
chloro
iodo
SMILES string
Clc1cc(Cl)cc(I)c1
InChI
1S/C6H3Cl2I/c7-4-1-5(8)3-6(9)2-4/h1-3H
InChI key
AATPRMRVLQZEHB-UHFFFAOYSA-N
Related Categories
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service