Skip to Content
Merck
All Photos(1)

Key Documents

222135

Sigma-Aldrich

4-Pentylaniline

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€65.90

€65.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€65.90

About This Item

Linear Formula:
CH3(CH2)4C6H4NH2
CAS Number:
Molecular Weight:
163.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€65.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.53 (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CCCCCc1ccc(N)cc1

InChI

1S/C11H17N/c1-2-3-4-5-10-6-8-11(12)9-7-10/h6-9H,2-5,12H2,1H3

InChI key

DGFTWBUZRHAHTH-UHFFFAOYSA-N

Application

4-Pentylaniline was used in the synthesis of 4-[(4′-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photo-crosslinkable liquid-crystalline azo-polymer for surface relief gratings and persistent fixation.
New. J. Chem., 33(6), 1343-1348 (2009)
László Szabó et al.
ChemistryOpen, 7(9), 720-729 (2018-09-28)
Given our possible future dependence on carbon fiber reinforced composites, the introduction of a renewable matrix might be advantageous for the vision of a sustainable world. Cellulose is a superior green candidate and provides exceptional freedom in composite design as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service