Skip to Content
Merck
All Photos(1)

Key Documents

23215

Sigma-Aldrich

1-Naphthyl chloroformate

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7ClO2
CAS Number:
Molecular Weight:
206.63
Beilstein:
512216
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

≥98.0% (GC)

refractive index

n20/D 1.598

density

1.273 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)Oc1cccc2ccccc12

InChI

1S/C11H7ClO2/c12-11(13)14-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

JRUFQZPDRRGHEF-UHFFFAOYSA-N

Application

1-Naphthyl chloroformate was used in preparation of:
  • 4-benzyl-2-(4-methoxyphenyl)oxazol-5-yl naphthalen-1-ylcarbonate[1]
  • naphthalen-1-yl 4-benzyl-2-(4-methoxyphenyl)-5-oxo-4,5-dihydrooxazole-4-carboxylate[1]
  • 2-(4-methoxyphenyl)-4-methyloxazol-5-yl naphthalen-1-ylcarbonate[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig D Campbell et al.
Organic & biomolecular chemistry, 9(11), 4205-4218 (2011-04-21)
The O- to C-carboxyl transfer of oxazolyl carbonates promoted by triazolinylidenes, generated in situ with NEt(3), shows a markedly different rate and chemoselectivity profile to the same reaction promoted by triazolinylidenes generated using KHMDS. The mechanism of these pathways has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service