Skip to Content
Merck
All Photos(1)

Key Documents

219924

Sigma-Aldrich

4-Hydroxy-3-nitrophenylacetic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€44.60

€44.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€44.60

About This Item

Linear Formula:
HOC6H3(NO2)CH2CO2H
CAS Number:
Molecular Weight:
197.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€44.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

146-148 °C (lit.)

solubility

ethanol: soluble 5%, clear, yellow to orange

functional group

carboxylic acid
nitro

SMILES string

OC(=O)Cc1ccc(O)c(c1)[N+]([O-])=O

InChI

1S/C8H7NO5/c10-7-2-1-5(4-8(11)12)3-6(7)9(13)14/h1-3,10H,4H2,(H,11,12)

InChI key

QBHBHOSRLDPIHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxy-3-nitrophenylacetic acid enhances the ice-nucleation activity of Xanthomonas campestris[1].

Application

4-Hydroxy-3-nitrophenylacetic acid was used in the synthesis of 4-hydroxy-3-nitrophenylacetyl caproic acid[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masayuki Oda et al.
Molecular immunology, 47(2-3), 357-364 (2009-10-06)
The effect of antibody affinity on molecular forms of immune complexes was investigated by measuring antigen-antibody interactions using surface plasmon resonance (SPR), electrospray ionization time-of-flight mass spectrometry under non-denaturing conditions (MS), analytical ultracentrifugation (AUC), and transmission electron microscopy (TEM). (4-Hydroxy-3-nitrophenyl)acetic
Matthew D Taylor et al.
Frontiers in immunology, 11, 1946-1946 (2020-09-10)
Recent studies have demonstrated that induction of a diverse repertoire of memory T cells ("immune education") affects responses to murine cecal ligation and puncture (CLP), the most widely - used animal model of sepsis. Among the documented effects of immune
P Bergqvist et al.
Mucosal immunology, 6(1), 122-135 (2012-07-13)
Whereas gut IgA responses to the microbiota may be multi-centered and diverse, little is known about IgA responses to T-cell-dependent antigens following oral immunizations. Using a novel approach, gut IgA responses to oral hapten (4-hydroxy-3-nitrophenyl)acetyl-cholera toxin (NP-CT) conjugates were followed
Gijs Hardenberg et al.
Immunology and cell biology, 86(6), 530-534 (2008-03-26)
The tumour necrosis factor (TNF) family member a proliferation-inducing ligand (APRIL) is implicated in various B-cell processes, such as class switch recombination, plasma cell differentiation and plasma cell survival. This was suggested from initial studies analysing B-cell responses in APRIL-deficient
H Torigoe et al.
The Journal of biological chemistry, 270(38), 22218-22222 (1995-09-22)
To understand the mechanism of affinity maturation, we examined the antigen-antibody interactions between 4-hydroxy-3-nitrophenylacetyl (NP) caproic acid and the Fab fragments of three anti-NP antibodies, N1G9, 3B44, and 3B62, by isothermal titration calorimetry. The analyses have revealed that all of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service